3,4-Dioxonaftaleno-1-sulfonato de sodio ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate
N0043-1G
N0043-25G
TCIAN0043-1GEA
16
EUR
TCIAN0043-1G
TCIAN0043-25G
3,4-Dioxonaftaleno-1-sulfonato de sodio ≥98.0% (by titrimetric analysis)
3,4-Dioxonaftaleno-1-sulfonato de sodio
Formula:
C₁₀H₅NaO₅S MW: 260.20 g/mol |
Número MDL:
MFCD00001700 Núm. CAS: 521-24-4 EINECS: 208-308-9 Índice Merck: 14,08646 |
|
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...