3-Aminocrotónato de metilo ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate

A1001-25G A1001-500G
TCIAA1001-25GEA 28 EUR
TCIAA1001-25G TCIAA1001-500G
3-Aminocrotónato de metilo ≥98.0% (by titrimetric analysis)
Methyl 3-aminocrotonate
Formula: C₅H₉NO₂
MW: 115,13 g/mol
Número MDL: MFCD00008072
Núm. CAS: 14205-39-1
EINECS: 238-056-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4247dcf644093ddff99448715c4fef25b7b0aba7&vid=4247dcf644093ddff99448715c4fef25b7b0aba7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR