CHROMABOND® PS SPE Columns
Proveedor: MACHEREY-NAGEL
731860 731861 731865 731866 731867 731868 731870 731871 731875 731877 730377 730378 730690 730310 731863 731909 730344 730376 730394 731862
MANA731860EA 252 EUR
MANA731860 MANA731861 MANA731865 MANA731866 MANA731867 MANA731868 MANA731870 MANA731871 MANA731875 MANA731877 MANA730377 MANA730378 MANA730690 MANA730310 MANA731863 MANA731909 MANA730344 MANA730376 MANA730394 MANA731862
CHROMABOND® PS SPE Columns
Columnas de cromatografía
Polymer-based SPE phases for RP and ion chromatography.
  • Base material: High purity polystyrene-divinylbenzene copolymers (PS/DVB)
  • Very low degree of swelling, thus very well suited for chromatography
  • Reliable application over the whole pH range from 0 to 14
CHROMABOND® PS columns and cartridges improve the chromatographic separation by removal of interfering components, which overlap with the analyte in the chromatogram. Furthermore, they extend the lifetime of the chromatographic column, since impurities can irreversibly block the column packing.