Capillary columns, Select FAME
Proveedor: VARIAN
CP7419 CP7419I5 CP7420 CP7421
VARICP7419EA 2400 EUR
VARICP7419 VARICP7419I5 VARICP7420 VARICP7421
Capillary columns, Select FAME
Columnas de cromatografía
The Select FAME column is tuned for optimal cis-trans separations of FAMEs, especially C₁₈ isomers.
  • Long lifetime due to high polarity 100% bonded phase
  • Low bleed provides more sensitivity for better detection limits
  • Better separation due to high efficiency and loadability for more accurate results
The Select FAME column also offers three times greater loadability, further improving the shape and separation for FAME isomers – especially if one component is present at a higher concentration.