Safety shoes, lace-up, Jalmatch
Proveedor: Jallatte
J0351/44 J0351/42 J0351/41 J0351/43 J0351/45 J0351/46 J0351/37 J0351/39 J0351/36 J0351/40 J0351/47 J0351/38
JALCJ0351/44EA 96.8 EUR
JALCJ0351/44 JALCJ0351/42 JALCJ0351/41 JALCJ0351/43 JALCJ0351/45 JALCJ0351/46 JALCJ0351/37 JALCJ0351/39 JALCJ0351/36 JALCJ0351/40 JALCJ0351/47 JALCJ0351/38
Safety shoes, lace-up, Jalmatch
Zapatos y botas
Laced safety shoes made from perforated suede leather, Softane™ dual-density PU sole (resistant to abrasion, hydrocarbons, oils and grease); with padded collar.
  • Lining with three dimensional foam structures for excellent ventilation
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
  • Perforated steel toecap (200 J)
Colour: Grey
Certificaciones: EN 345-1; S1P