Nitrile gloves, Showa 707FL
Proveedor: Showa
707FL-06 707FL-07 707FL-10 707FL-08 707FL-11 707FL-09
112-2748EA 33 EUR
112-2748 112-2749 112-2775 112-2773 112-2776 112-2774
Nitrile gloves, Showa 707FL
Guantes
These lined nitrile gloves with rolled cuffs protect the hand from oils, hydrocarbons, grease and abrasion.
  • Non sterile, powder-free, smooth finish
  • Comfortable flock lining for continuous wear
  • Impermeable for working in damp or greasy environments
  • Tractor tread finish and 'slip-on' treatment provide good grip
  • Easy to put on and remove
  • Semi-disposable; can be used once or re-used
Suitable for manufacturing, laboratories, food processing, light duty maintenance, clean up tasks, paint applications, fuel handling and refineries.
Certificaciones: EN ISOMore Product Information