SOLA™ SPE cartridges
Proveedor: Thermo Fisher Scientific
60109-003 60109-002 60109-001 60109-004 60109-005
554-1985EA 252 EUR
554-1985 554-1984 554-1983 554-4393 554-4394
SOLA™ SPE cartridges
Columnas de cromatografía
This fritless SPE product range provides greater reproducibility with cleaner, more consistent extracts. SOLA™ products provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products. This includes:
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Increased sensitivity
This increased performance gives higher confidence in analytical results without compromising ease of use or requiring complex method development.
SOLA™ products provide reduced failure rates, higher analysis speeds and lower solvent requirements.
SOLA™ products provide reduced failure rates, higher analysis speeds and lower solvent requirements.