4-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Chloro-2-methylbenzeneboronic acid
C2577-1G
C2577-5G
TCIAC2577-1GEA
35
EUR
TCIAC2577-1G
TCIAC2577-5G
4-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-methylphenylboronic acid
Formula:
C₇H₈BClO₂ MW: 170,40 g/mol |
Número MDL:
MFCD02683107 Núm. CAS: 209919-30-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...