Dibenzyl phosphate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

P1120-25G P1120-5G
TCIAP1120-25GEA 246 EUR
TCIAP1120-25G TCIAP1120-5G
Dibenzyl phosphate ≥98.0% (by HPLC, titration analysis)
Dibenzyl phosphate
Formula: C₁₄H₁₅O₄P
Temperatura de almacenaje: Congelador
Número MDL: MFCD00004775
Núm. CAS: 1623-08-1
EINECS: 216-602-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12ef3b4f7f53df5cd4638d2df6c8612bf69476b5&vid=12ef3b4f7f53df5cd4638d2df6c8612bf69476b5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 82.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR