2-Mercapto-6-nitrobenzothiazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Nitrobenzo[d]thiazole-2(3H)-thione
N0326-25G
N0326-5G
TCIAN0326-25GEA
250
EUR
TCIAN0326-25G
TCIAN0326-5G
2-Mercapto-6-nitrobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Mercapto-6-nitrobenzothiazole
Formula:
C₇H₄N₂O₂S₂ MW: 212,24 g/mol |
Número MDL:
MFCD00041850 Núm. CAS: 4845-58-3 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...