2,2'-Dibenzothiazole disulphide ≥96.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,2'-Dithiobisbenzothiazole, 2,2-Dithiobis(benzothiazole), 2,2'-Dibenzothiazolyl disulphide
D0538-25G
D0538-500G
TCIAD0538-25GEA
17
EUR
TCIAD0538-25G
TCIAD0538-500G
2,2'-Dibenzothiazole disulphide ≥96.0% (by HPLC, total nitrogen)
2,2'-Dibenzothiazole disulphide
Formula:
C₁₄H₈N₂S₄ MW: 332,4867 g/mol |
Número MDL:
MFCD00022874 Núm. CAS: 120-78-5 EINECS: 204-424-9 UN: 3077 ADR: 9,III Índice Merck: 14,03370 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...