3-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid
C2028-1G
C2028-5G
TCIAC2028-1GEA
36
EUR
TCIAC2028-1G
TCIAC2028-5G
3-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
3-Carboxyphenylboronic acid
Formula:
C₇H₇BO₄ MW: 165,94 g/mol Pto. de fusión: 240°C |
Número MDL:
MFCD00036833 Núm. CAS: 25487-66-5 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...