3-Aminophenylboronic acid hemisulphate
Proveedor: TCI
|
Atención
|
A1774-1G
A1774-25G
A1774-5G
TCIAA1774-1GEA
29
EUR
TCIAA1774-1G
TCIAA1774-25G
TCIAA1774-5G
3-Aminophenylboronic acid hemisulphate
3-Aminophenylboronic acid hemisulphate
Formula:
C₁₂H₁₈B₂N₂O₈S |
Número MDL:
MFCD00013111 Núm. CAS: 66472-86-4 EINECS: 266-376-5 |
|
Resultados test especificaciones
Appearance | White to Amber to Dark green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...