2-Amino-4-chloro-5-nitrophenol ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

A1228-25G A1228-250G
TCIAA1228-25GEA 47 EUR
TCIAA1228-25G TCIAA1228-250G
2-Amino-4-chloro-5-nitrophenol ≥98.0% (by titrimetric analysis)
2-Amino-4-chloro-5-nitrophenol
Formula: C₆H₅ClN₂O₃
Número MDL: MFCD00010300
Núm. CAS: 6358-07-2
EINECS: 228-760-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59d122704058b0cfb8e04d66790e16e41ded600&vid=f59d122704058b0cfb8e04d66790e16e41ded600&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 219.0 to 222.0 deg-C

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR