N,N-Bis(trifluoromethanesulphonyl)aniline ≥98.0% (por GC) triflating reagent
Proveedor: TCI
|
Atención
|
P1257-25G
P1257-5G
TCIAP1257-25GEA
154
EUR
TCIAP1257-25G
TCIAP1257-5G
N,N-Bis(trifluoromethanesulphonyl)aniline ≥98.0% (por GC) triflating reagent
N,N-Bis(trifluoromethanesulphonyl)aniline
Formula:
C₈H₅F₆NO₄S₂ |
Número MDL:
MFCD00000404 Núm. CAS: 37595-74-7 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 100.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...