3'-Bromo-5'-chloro-2'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(3-Bromo-5-chloro-2-hydroxyphenyl)ethanone, 5-Chloro-3-bromo-2-hydroxyacetophenone
B2236-25G
B2236-5G
TCIAB2236-25GEA
248
EUR
TCIAB2236-25G
TCIAB2236-5G
3'-Bromo-5'-chloro-2'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
Formula:
C₈H₆BrClO₂ MW: 249,49 g/mol |
Número MDL:
MFCD00857068 Núm. CAS: 59443-15-1 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 98.0 to 102.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...