2'-Amino-3'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(2-Amino-3-hydroxyphenyl)ethanone
A1321-1G
A1321-5G
TCIAA1321-1GEA
111
EUR
TCIAA1321-1G
TCIAA1321-5G
2'-Amino-3'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Amino-3'-hydroxyacetophenone
Formula:
C₈H₉NO₂ MW: 151,1626 g/mol |
Número MDL:
MFCD00191301 Núm. CAS: 4502-10-7 |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...