2-Biphenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

B2488-25G B2488-5G
TCIAB2488-25GEA 318 EUR
TCIAB2488-25G TCIAB2488-5G
2-Biphenylboronic acid (contains varying amounts of Anhydride)
2-Biphenylboronic acid
Formula: C₁₂H₁₁BO₂
MW: 198,03 g/mol
Pto. de fusión: 129°C
Número MDL: MFCD00136929
Núm. CAS: 4688-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f1976d8d5ae6b9b2de49f6c6648ba550c89cfb7&vid=3f1976d8d5ae6b9b2de49f6c6648ba550c89cfb7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR