3-Biphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
[1,1'-Biphenyl]-3-ylboronic acid
B2489-25G
B2489-5G
TCIAB2489-25GEA
236
EUR
TCIAB2489-25G
TCIAB2489-5G
3-Biphenylboronic acid (contains varying amounts of Anhydride)
3-Biphenylboronic acid
Formula:
C₁₂H₁₁BO₂ MW: 198,03 g/mol Pto. de fusión: 208°C |
Número MDL:
MFCD01318102 Núm. CAS: 5122-95-2 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...