4-Amino-2,6-diphenylphenol ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

A1269-1G A1269-500MG
TCIAA1269-1GEA 126 EUR
TCIAA1269-1G TCIAA1269-500MG
4-Amino-2,6-diphenylphenol ≥98.0% (by titrimetric analysis)
4-Amino-2,6-diphenylphenol
Formula: C₁₈H₁₅NO
Número MDL: MFCD00034070
Núm. CAS: 50432-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33d2c8560cc2ab4fa63e955a3811f48bdb68e5c2&vid=33d2c8560cc2ab4fa63e955a3811f48bdb68e5c2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Brown to Dark purple powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR