2-Pyridinesulphonic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Pyridine-2-sulphonic acid
P1647-25G
P1647-5G
TCIAP1647-25GEA
284
EUR
TCIAP1647-25G
TCIAP1647-5G
2-Pyridinesulphonic acid ≥98.0% (by HPLC, titration analysis)
2-Pyridinesulphonic acid
Formula:
C₅H₅NO₃S MW: 159.1631 g/mol Pto. de fusión: 252°C |
Número MDL:
MFCD00234080 Núm. CAS: 15103-48-7 UN: 3261 ADR: 8 III |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...