Allylamine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Allylamine Hydrochloride

A0226-25G A0226-500G A0226-100G
TCIAA0226-25GEA 32 EUR
TCIAA0226-25G TCIAA0226-500G TCIAA0226-100G
Allylamine hydrochloride ≥98.0% (by titrimetric analysis)
Allylamine hydrochloride
Formula: C₃H₈ClN
MW: 93,5553 g/mol
Pto. de fusión: 106°C
Número MDL: MFCD00054329
Núm. CAS: 10017-11-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83c5a843151aed3b256a22212e9ee01403dbbb66&vid=83c5a843151aed3b256a22212e9ee01403dbbb66&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR