Sodium-1-undecanesulphonate, (max. 2% H₂O) 98+% (dry weight)
Proveedor: Thermo Fisher Scientific
B20332.14
B20332.22
B20332.14EA
211
EUR
B20332.14
B20332.22
Sodium-1-undecanesulphonate, (max. 2% H₂O) 98+% (dry weight)
Sodium-1-undecanesulphonate
Purity: 98+%
CAS No.: 5838-34-6
Molecular Formula: CH3(CH2)10SO3Na
Formula Weight: 258.36
Melting Point:
Formula:
C₁₂H₂₅NaO₃S Pto. de fusión: >300 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00053627 Núm. CAS: 5838-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...