4-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid
F0666-1G
F0666-5G
TCIAF0666-1GEA
46
EUR
TCIAF0666-1G
TCIAF0666-5G
4-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-2-methylphenylboronic acid
Formula:
C₇H₈BFO₂ MW: 153,95 g/mol |
Número MDL:
MFCD02093072 Núm. CAS: 139911-29-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...