2,3-Dimethyl-4-nitrophenol ≥98.0% (por GC)

Proveedor: TCI
Atención

D2608-25G D2608-5G
TCIAD2608-25GEA 307 EUR
TCIAD2608-25G TCIAD2608-5G
2,3-Dimethyl-4-nitrophenol ≥98.0% (por GC)
2,3-Dimethyl-4-nitrophenol
Formula: C₈H₉NO₃
MW: 167,1600 g/mol
Número MDL: MFCD01861284
Núm. CAS: 19499-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26b9ed79e244f23af59ada4c597957474e7b496&vid=c26b9ed79e244f23af59ada4c597957474e7b496&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 127.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR