3,5-Di-tert-butylbenzyl bromide ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
1-(Bromomethyl)-3,5-di-tert-butylbenzene
D2653-1G
D2653-5G
TCIAD2653-1GEA
50
EUR
TCIAD2653-1G
TCIAD2653-5G
3,5-Di-tert-butylbenzyl bromide ≥98.0% (por GC)
3,5-Di-tert-butylbenzyl bromide
Formula:
C₁₅H₂₃Br MW: 283,2471 g/mol |
Número MDL:
MFCD03701623 Núm. CAS: 62938-08-3 UN: 1759 ADR: 8,III |
Resultados test especificaciones
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 33.0 to 37.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...