trans-1,4-Cyclohexanedicarboxylic acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C0475-25G C0475-500G C0475-100G
TCIAC0475-25GEA 28 EUR
TCIAC0475-25G TCIAC0475-500G TCIAC0475-100G
trans-1,4-Cyclohexanedicarboxylic acid ≥97.0% (by GC, titration analysis)
trans-1,4-Cyclohexanedicarboxylic acid
Formula: C₈H₁₂O₄
MW: 172,18 g/mol
Número MDL: MFCD00066203
Núm. CAS: 619-82-9
EINECS: 210-614-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ef28fdbb866cdddf6c53d5762858378305e5130&vid=8ef28fdbb866cdddf6c53d5762858378305e5130&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR