Potassium hydrogen diiodate 99.95-100.05% ACS primary standard
Proveedor: Thermo Fisher Scientific
|
Peligro
|
41986.14
41986.22
41986.14EA
83.3
EUR
41986.14
41986.22
Potassium hydrogen diiodate 99.95-100.05% ACS primary standard
Potassium hydrogen diiodate
Formula:
HI₂KO₆ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011400 Núm. CAS: 13455-24-8 EINECS: 236-650-9 UN: 3085 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...