Tungsten boride ≥99% (metals basis)
Proveedor: Thermo Fisher Scientific
12559.14
12559.22
12559.14EA
143
EUR
12559.14
12559.22
Tungsten boride ≥99% (metals basis)
Tungsten boride
Purity: 99%
CAS No.: 12007-09-9
Molecular Formula: WB
Formula Weight: 194.66
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00049708
Formula:
BW Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00049708 Núm. CAS: 12007-09-9 EINECS: 234-498-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...