2,5-Dihidroxibencenosulfonato de potasio ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate

H0593-25G H0593-500G
TCIAH0593-25GEA 16 EUR
TCIAH0593-25G TCIAH0593-500G
2,5-Dihidroxibencenosulfonato de potasio ≥98.0% (by titrimetric analysis)
2,5-Dihidroxibencenosulfonato de potasio
Formula: C₆H₅KO₅S
Número MDL: MFCD00007475
Núm. CAS: 21799-87-1
EINECS: 244-584-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86a297df1dff02921ed92e7eb18637cb1f4b7c9&vid=e86a297df1dff02921ed92e7eb18637cb1f4b7c9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Cerium redox Method) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR