Tetraciclina

Proveedor: Thermo Scientific
Atención

J61714.06 J61714.14 J61714.22
J61714.06EA 43.1 EUR
J61714.06 J61714.14 J61714.22
Tetraciclina
Tetraciclina

CAS No.: 60-54-8 Molecular Formula: C22H24N2O8 Formula Weight: 444.43 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: Pale yellow Melting Point: 172-174° Solubility: Limited solubility in water. Soluble in 1M HCl with heating. Beilstein Reference: 2230417 Merck Reference: 14,9196 MDL No.: MFCD00151232


Can induce apoptosis in osteoclasts

Formula: C₂₂H₂₄N₂O₈
MW: 444,43 g/mol
Pto. de fusión: 172…174 °C
Número MDL: MFCD00151232
Núm. CAS: 60-54-8
EINECS: 200-481-9
Índice Merck: 14,9196

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR