Eritromicina cell culture reagent

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: (-)-Erythromycin

J62279.14 J62279.09
J62279.14EA 210 EUR
J62279.14 J62279.09
Eritromicina cell culture reagent
Eritromicina

CAS No.: 114-07-8 Molecular Formula: C37H67NO13 Formula Weight: 733.93 Physical Form: Crystalline powder Appearance: White Solubility: Soluble in water at 2mg/ml Beilstein Reference: 75279 Merck Reference: 14,3681 MDL No.: MFCD00084654


Macrolide antibiotic that inhibits bacterial protein synthesis

Formula: C₃₇H₆₇NO₁₃
MW: 733.95 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00084654
Núm. CAS: 114-07-8
EINECS: 204-040-1
Índice Merck: 14,3681

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR