Puromicina diclorhidrato ≥99%
Proveedor: Thermo Fisher Scientific
|
Atención
|
Purity: 99%
CAS No.: 58-58-2
Molecular Formula: C22H29N7O5.2HCl
Formula Weight: 544.44
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 168-170°
Solubility: Soluble in water
Beilstein Reference: 3853613
Merck Reference: 14,7943
MDL No.: MFCD00012691
Causes premature chain termination in protein synthesis
Formula:
C₂₂H₂₉N₇O₅·2HCl Pto. de fusión: 168…170 °C Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00012691 Núm. CAS: 58-58-2 EINECS: 200-387-8 Índice Merck: 14,7943 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...