Mini desiccator cabinets, SICCO, Premium
Proveedor: Bohlender
V1950-01 V1950-02 V1950-03
467-0130EA 189 EUR
467-0130 467-0131 467-0132
Mini desiccator cabinets, SICCO, Premium
Desecadores Armarios desecadores
Clear polycarbonate. Each chamber has a single door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight and portable
  • Choice of units with one, two or three chambers, each chamber supplied with two shelves
  • Supplied with hygrometer and desiccant which is in a handy resealable container