HPLC columns, Hypersil GOLD™
Proveedor: Thermo Fisher Scientific
25005-014006 25002-012135 25005-012106 25002-104630 25003-012106 25003-254630 25005-304030 25005-059070A 25005-059270A 25005-109070A 25005-109270A 25005-159070A 25005-159270A 25005-159370A 25005-259070A 25005-259270A 25005-259370A 25012-159270A 25012-159370A 25012-259070A 25012-259270A 25012-259370A 25012-259570A
555-8052EA 580 EUR
555-8052 555-8133 555-8050 555-8002 555-8007 555-8048 555-7695 555-0140 555-0141 555-0142 555-0143 555-0144 555-0145 555-0146 555-0147 555-0148 555-0149 555-0150 555-0151 555-0152 555-0153 555-0154 555-0155
HPLC columns, Hypersil GOLD™
Columnas de cromatografía
These Hypersil GOLD™ HPLC columns are ideal for LS/MS applications to achieve exceptional peak shape and resolution.
  • Endcapped and ultrapure
  • Silica-based columns with exceptional peak shape and resolution
  • Rugged sleeved design for robustness
  • High performance and sensitivity
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality ofMore Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Carga de carbono 10% 10% 10%
Rango de pH 1 - 11 1 to 11 1 - 11
Surface area 220 m²/g 220 m²/g 220 m²/g