3-(N,N-Dimethylamino)phenylboronic acid hydrochloride (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

D4013-1G D4013-5G
TCIAD4013-1GEA 68 EUR
TCIAD4013-1G TCIAD4013-5G
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride (contains varying amounts of Anhydride)
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride
Formula: C₈H₁₃BClNO₂
MW: 201,4583 g/mol
Número MDL: MFCD04112542
Núm. CAS: 1256355-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da78ce2f0243b368003515025d4d053c635ddc37&vid=da78ce2f0243b368003515025d4d053c635ddc37&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) 98.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR