(-)-Menthyl chloroformate ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
(1R)-(-)-Menthyl chloroformate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl carbonochloridate
M0990-25ML
M0990-5ML
TCIAM0990-25MLEA
140
EUR
TCIAM0990-25ML
TCIAM0990-5ML
(-)-Menthyl chloroformate ≥97.0% (by titrimetric analysis)
(-)-Menthyl chloroformate
Formula:
C₁₁H₁₉ClO₂ MW: 218.73 g/mol Pto. de inflamación: 70°C |
Número MDL:
MFCD00009694 Núm. CAS: 14602-86-9 UN: 3277 ADR: 6.1 II |
|
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (Argentmetric Titration) | min. 97.0 % |
Specific rotation [a]20/D | -75.0 to -80.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...