(+)-N,N,N',N'-Tetramethyl-L-tartardiamide ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: (2R,3R)-N,N,N',N'-Tetramethyl-L-tartaric acid diamide

T1839-25G T1839-5G
TCIAT1839-25GEA 178 EUR
TCIAT1839-25G TCIAT1839-5G
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide ≥98.0% (por GC)
(+)-N,N,N',N'-Tetramethyl-L-tartardiamide
Formula: C₈H₁₆N₂O₄
MW: 204,2236 g/mol
Número MDL: MFCD00025672
Núm. CAS: 26549-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad2be9ed4a0fb82de2f15dc3f07d9812a4693bd7&vid=ad2be9ed4a0fb82de2f15dc3f07d9812a4693bd7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 185.0 to 189.0 deg-C
Specific rotation [a]20/D +43.0 to +48.0 deg (C=1,EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR