(-)-N,N,N',N'-Tetramethyl-D-tartardiamide ≥98.0% (by GC, total nitrogen)

Proveedor: TCI

Sinónimos: (2S,3S)-2,3-Dihydroxy-N1,N1,N4,N4-tetramethylsuccinamide

T1864-25G T1864-5G
TCIAT1864-25GEA 318 EUR
TCIAT1864-25G TCIAT1864-5G
(-)-N,N,N',N'-Tetramethyl-D-tartardiamide ≥98.0% (by GC, total nitrogen)
(-)-N,N,N',N'-Tetramethyl-D-tartardiamide
Formula: C₈H₁₆N₂O₄
MW: 204,2236 g/mol
Número MDL: MFCD00077879
Núm. CAS: 63126-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3078e1ae01da78b6b181a4fd84343f86e8f4b95&vid=e3078e1ae01da78b6b181a4fd84343f86e8f4b95&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 186.0 to 190.0 deg-C
Specific rotation [a]20/D -43.0 to -49.0 deg (C=3, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR