N-Phenyliminodiacetic acid 97%
Proveedor: Thermo Fisher Scientific
|
Atención
|
L08437.06
L08437.14
L08437.06EA
72.1
EUR
L08437.06
L08437.14
N-Phenyliminodiacetic acid 97%
N-Phenyliminodiacetic acid
Purity: 97%
CAS No.: 1137-73-1
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Melting Point: ca 140° dec.
Beilstein Reference: 2808435
Formula:
C₁₀H₁₁NO₄ Pto. de fusión: ∼ 140 °C (desc.) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00043570 Núm. CAS: 1137-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...