trans-3-Chloroacrylic acid ≥99%
Proveedor: Thermo Scientific
|
Peligro
|
A15527.03
A15527.06
A15527.14
A15527.MD
A15527.03EA
95.2
EUR
A15527.03
A15527.06
A15527.14
A15527.MD
trans-3-Chloroacrylic acid ≥99%
trans-3-Chloroacrylic acid
Purity: 99%
CAS No.: 2345-61-1
Molecular Formula: C3H3ClO2
Formula Weight: 106.51
Storage Temperature: 2°C to 8°C
Melting Point: 81-85°
Beilstein Reference: 1720279
UN No.: 3261
MDL No.: MFCD00064237
Formula:
C₃H₃ClO₂ MW: 106,51 g/mol Pto. de fusión: 81…85 °C |
Número MDL:
MFCD00064237 Núm. CAS: 2345-61-1 EINECS: 219-070-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...