Zinc (II) phthalocyanine (Sulphate ionophore II) ≥95.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex
P0767-25G
P0767-1G
P0767-5G
TCIAP0767-25GEA
164
EUR
TCIAP0767-25G
TCIAP0767-1G
TCIAP0767-5G
Zinc (II) phthalocyanine (Sulphate ionophore II) ≥95.0% (by titrimetric analysis)
Zinc (II) phthalocyanine (Sulphate ionophore II)
Formula:
C₃₂H₁₆N₈Zn MW: 577.91 g/mol |
Número MDL:
MFCD00041955 Núm. CAS: 14320-04-8 EINECS: 238-262-5 |
|
Resultados test especificaciones
Appearance | Dark red to Dark purple to Dark blue powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Lambda max. | 668.0 to 670.0 nm (DMF) |
Molar extinction coefficient | min. 140,000 (668.0 to 670.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...