4-Fluoro-4'-methylchalcone
Proveedor: TCI
|
Atención
|
F0460-25G
F0460-5G
TCIAF0460-25GEA
124
EUR
TCIAF0460-25G
TCIAF0460-5G
4-Fluoro-4'-methylchalcone
4-Fluoro-4'-methylchalcone
Formula:
C₁₆H₁₃FO |
Número MDL:
MFCD00017981 Núm. CAS: 13565-38-3 |
|
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...