D-Glucoronic acid ≥96.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

G0302-25G G0302-5G
TCIAG0302-25GEA 112 EUR
TCIAG0302-25G TCIAG0302-5G
D-Glucoronic acid ≥96.0% (by HPLC, titration analysis)
D-Glucoronic acid
Formula: C₆H₁₀O₇
MW: 194.139 g/mol
Pto. de fusión: 153°C
Número MDL: MFCD00166981
Núm. CAS: 6556-12-3
EINECS: 229-486-4
Índice Merck: 14,04465

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D +34.0 ~ +38.0 deg (C=6, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR