Retinoic acid ≥93.0% (by HPLC)

Proveedor: TCI
Atención

Sinónimos: all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin

R0064-1G R0064-5G
TCIAR0064-1GEA 107 EUR
TCIAR0064-1G TCIAR0064-5G
Retinoic acid ≥93.0% (by HPLC)
Retinoic acid
Formula: C₂₀H₂₈O₂
Número MDL: MFCD00001551
Núm. CAS: 302-79-4
EINECS: 206-129-0
Índice Merck: 14,08165
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06b0c46d9e7ac7f95689098d502ba896c643e8f9&vid=06b0c46d9e7ac7f95689098d502ba896c643e8f9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR