3,4-(Methylenedioxy)phenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid
M2035-25G
M2035-5G
TCIAM2035-25GEA
250
EUR
TCIAM2035-25G
TCIAM2035-5G
3,4-(Methylenedioxy)phenylboronic acid (contains varying amounts of Anhydride)
3,4-(Methylenedioxy)phenylboronic acid
Formula:
C₇H₇BO₄ MW: 165,94 g/mol |
Número MDL:
MFCD01009695 Núm. CAS: 94839-07-3 |
Resultados test especificaciones
Appearance | White to Green to Brown powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...