(Cyanomethyl)tributylphosphonium chloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Tributyl(cyanomethyl)phosphonium chloride
C1442-10G
C1442-25G
TCIAC1442-10GEA
72
EUR
TCIAC1442-10G
TCIAC1442-25G
(Cyanomethyl)tributylphosphonium chloride ≥98.0% (by titrimetric analysis)
(Cyanomethyl)tributylphosphonium chloride
Formula:
C₁₄H₂₉ClNP MW: 277,8135 g/mol |
Número MDL:
MFCD00671508 Núm. CAS: 82358-61-0 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...