Diphenylphosphoryl azide ≥97.0% (por GC)
Proveedor: TCI
|
Peligro
|
D1672-25G
D1672-250G
D1672-5G
TCIAD1672-25GEA
45
EUR
TCIAD1672-25G
TCIAD1672-250G
TCIAD1672-5G
Diphenylphosphoryl azide ≥97.0% (por GC)
Diphenylphosphoryl azide
Formula:
C₁₂H₁₀N₃O₃P Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00001987 Núm. CAS: 26386-88-9 EINECS: 247-644-0 UN: 3278 ADR: 6.1,II |
|
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...