Trithiocyanuric acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Trimercaptotriazine

T0935-500G T0935-25G T0935-100G
TCIAT0935-500GEA 128 EUR
TCIAT0935-500G TCIAT0935-25G TCIAT0935-100G
Trithiocyanuric acid ≥98.0% (by titrimetric analysis)
Trithiocyanuric acid
Formula: C₃H₃N₃S₃
Número MDL: MFCD00006052
Núm. CAS: 638-16-4
EINECS: 211-322-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a30144203f1fb2bdae1fcfadcdc75d2cdb4a53e&vid=8a30144203f1fb2bdae1fcfadcdc75d2cdb4a53e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR