Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III) ≥97%
Proveedor: Thermo Fisher Scientific
|
Peligro
|
44381.03
44381.06
44381.03EA
162
EUR
44381.03
44381.06
Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III) ≥97%
Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III)
Purity: 97%
CAS No.: 175923-07-6
Molecular Formula: C18H54LaN3Si6
Formula Weight: 620.06
Flash Point: 2°(36°F)
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 149-152°
UN No.: 3396
MDL No.: MFCD00271009
Formula:
C₁₈H₅₄LaN₃Si₆ Pto. de fusión: 149…152 °C Pto. de inflamación: 2 °C (36 °F) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00271009 Núm. CAS: 175923-07-6 UN: 3396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...